Learn More
Bon Opus Biosciences Recombinant Mouse SIGLEC-15 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP106250UG
Encompass_Preferred
Recombinant Mouse SIGLEC-15 (C-hFc) is produced by Human Cells. The target gene encoding Arg24-Thr262 is expressed with a C-hFc tag. Accession# Q6ZMC9-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1062
Catalog No. 50-217-2660