Learn More
Bon Opus Biosciences Recombinant Mouse SIGLEC-15 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP106150UG
Encompass_Preferred
Recombinant Mouse SIGLEC-15 (C-His) is produced by Human Cells. The target gene encoding Arg24-Thr262 is expressed with a C-His tag. Accession# Q6ZMC9-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1061
Catalog No. 50-217-2659