Learn More
Sino Biological Recombinant Mouse Siglec-15 Protein (rFc Tag)
Supplier:  Sino Biological 5A4074M31H200
Encompass_Preferred
A DNA sequence encoding the mouse SIGLEC15 (NP_003865.1) (Met1-Gly225) was expressed with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-255-2936