Learn More
BIOHIPPO Mouse sIL-6R Microsample
SDP
Catalog No. 5000315047
Encompass
Mouse sIL-6R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sIL-6R in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 118 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse sIL-6R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse sIL-6R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse sIL-6R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15047 Supplier BIOHIPPO Supplier No. BHE15211629
Mouse sIL-6R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sIL-6R in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 118 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse sIL-6R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse sIL-6R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse sIL-6R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color