Learn More
ACROBiosystems Mouse SIRP alpha 25ug
Catalog No. 502534175
Encompass_Preferred
Biotinylated Mouse SIRP alpha Protein FcAvitag (SIA-M82F7) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # P97797-1).
Catalog No. 50-253-4175 Supplier ACROBiosystems Supplier No. SIAM82F725UG
Biotinylated Mouse SIRP alpha Protein FcAvitag (SIA-M82F7) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # P97797-1).