Learn More
ACROBiosystems ACROBiosystems Mouse SIRP alpha / CD172a Protein, His Tag
Catalog No. 502106690
Encompass_Preferred
Recombinant Protein;1MG;Mouse SIRP alpha, His Tag (SIA-M52H4) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # BAA13520.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.8 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-M525a) at 2 μg/mL (100 μL/well) can bind Mouse SIRP alpha, His Tag (Cat. No. SIA-M52H4) with a linear range of 0.02-0.625 μg/mL (QC tested).
Catalog No. 50-210-6690 Supplier ACROBiosystems Supplier No. SIAM52H41MG
Recombinant Protein;1MG;Mouse SIRP alpha, His Tag (SIA-M52H4) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # BAA13520.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.8 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-M525a) at 2 μg/mL (100 μL/well) can bind Mouse SIRP alpha, His Tag (Cat. No. SIA-M52H4) with a linear range of 0.02-0.625 μg/mL (QC tested).