Learn More
ACROBiosystems ACROBiosystems Mouse SIRP alpha / CD172a Protein, Mouse IgG2a Fc Tag, low endotoxin
Supplier:  ACROBiosystems SIAM5252100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (SIA-M5252) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # BAA13520.1 (G365D)).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 65.0 kDa. The protein migrates as 90-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag (Cat. No. SIA-M5252) at 2 μg/mL (100 μL/well) can bind Mouse CD47, Fc Tag (Cat. No. CD7-M5251) with a linear range of 8-63 ng/mL (QC tested).;Immobilized Human CD47 at 10 μg/mL (100 μL/well) can bind Mouse SIRP alpha, Mouse IgG2a Fc Tag (Cat. No. SIA-M5252) with a linear range of 0.02-0.313 μg/mL (Routinely tested).
Catalog No. 50-210-6729