Learn More
Sino Biological Recombinant Anti-SIRP alpha/SHPS-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50956R00120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIRP alpha / CD172a (Catalog#50956-M08H; BAA20376.1; Met1-Asn373).
Catalog No. 50-241-0895