Learn More
ACROBiosystems ACROBiosystems Mouse SLAMF7 / CRACC / CD319 Protein, Mouse IgG2a Fc Tag, low endotoxin
Catalog No. 502106732
Encompass_Preferred
Recombinant Protein;100UG;Mouse SLAMF7, Mouse IgG2a Fc Tag, low endotoxin (SL7-M5252) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 224 (Accession # Q8BHK6-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 49.4 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6732 Supplier ACROBiosystems Supplier No. SL7M5252100UG
Recombinant Protein;100UG;Mouse SLAMF7, Mouse IgG2a Fc Tag, low endotoxin (SL7-M5252) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Gly 224 (Accession # Q8BHK6-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 49.4 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.