Learn More
Sino Biological Anti-SLAMF7/CRACC/CS1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50201T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SLAMF7/CRACC/CS1 (rM SLAMF7/CRACC/CS1; Catalog#50201-M08H; NP_653122.2; Met1-Gly224). SLAMF7/CRACC/CS1 specific IgG was purified by Mouse SLAMF7/CRACC/CS1 affinity chromatography.
Catalog No. 50-255-3381