Learn More
Sino Biological Anti-SLAMF7/CRACC/CS1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50201RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SLAMF7 extracellular domain (rM SLAMF7; Catalog#50201-M08H; NP_653122.2; Met 1-Gly 224). SLAMF7 specific IgG was purified by mouse SLAMF7 affinity chromatography.
Catalog No. 50-240-8105