Learn More
Sino Biological Mouse SLAMF8 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 100358T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SLAMF8 (Catalog#50623-M08H; NP_083360.2; Met 1-Asp 231). SLAMF8 specific IgG was purified by Mouse SLAMF8 affinity chromatography.
Catalog No. 50-237-4019