Learn More
Sino Biological Recombinant Mouse CD98 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50813M07H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse SLC3A2 (NP_001154885.1) (Ala 139-Ala 565) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-3856