Learn More
Biomatik Corporation Mouse Slit Homolog 1 (Slit1) ELISA Kit
Supplier:  Biomatik Corporation EKU07366
Encompass_Preferred
Mouse Slit Homolog 1 (Slit1) ELISA Kit for detecting Slit1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q80TR4. Gene ID: 20562. Target Synonyms: MEGF4; SLIL1; SLIT3; Multiple epidermal growth factor-like domains protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6945