Learn More
Biomatik Corporation Mouse Small Ubiquitin Related Modifier Protein 1 (SUMO1) ELISA Kit
Supplier:  Biomatik Corporation EKU08390
Encompass_Preferred
Mouse Small Ubiquitin Related Modifier Protein 1 (SUMO1) ELISA Kit for detecting SUMO1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63166. Gene ID: 22218. Target Synonyms: GMP1; OFC10; PIC1; SMT3; SMT3C; SMT3H3; SUMO-1; UBL1; GAP-modifying protein 1; Ubiquitin-homology domain protein PIC1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8671