Learn More
Sino Biological Recombinant Mouse alpha-Synuclein Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 57031M08E100
Encompass_Preferred
A DNA sequence encoding the Mouse SNCA (O55042-1) (Met1-Ala140) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-7663