Learn More
American Research Products Inc Mouse Soluable Thrombomodulin
Catalog No. 5000329470
Encompass
Mouse Soluable Thrombomodulin (STM) ELISA KIT from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29470 Supplier American Research Products Inc Supplier No. CSBE13383M
Mouse Soluable Thrombomodulin (STM) ELISA KIT from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich