Learn More
American Research Products Inc Mouse Soluble Endoglin ENG sC
Catalog No. 5000326355
Encompass
Mouse Soluble Endoglin ENG/sCD105 ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26355 Supplier American Research Products Inc Supplier No. CSBE09921M
Mouse Soluble Endoglin ENG/sCD105 ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Sandwich