Learn More
American Research Products Inc Mouse soluble interleukin 2 re
Catalog No. 5000328830
Encompass
Mouse soluble interleukin-2 receptor sIL-2Ra ELISA kit from CUSABIO Detection Range 15 6 pg/ml - 1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28830 Supplier American Research Products Inc Supplier No. CSBE07440M
Mouse soluble interleukin-2 receptor sIL-2Ra ELISA kit from CUSABIO Detection Range 15 6 pg/ml - 1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich