Learn More
American Research Products Inc Mouse soluble Lectin like OXid
Catalog No. 5000326710
Encompass
Mouse soluble Lectin-like OXidized Low Density Lipoprotein Receptor-1 (sLOX-1) ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26710 Supplier American Research Products Inc Supplier No. CSBE15846M
Mouse soluble Lectin-like OXidized Low Density Lipoprotein Receptor-1 (sLOX-1) ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich