Learn More
American Research Products Inc Mouse soluble receptor actiVat
Catalog No. 5000328805
Encompass
Mouse soluble receptor actiVator of nuclear factor-kB ligand sRANKL ELISA Kit from CUSABIO Detection Range 12 5 pg/ml-800 pg/ml Sensitivity 3 12 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28805 Supplier American Research Products Inc Supplier No. CSBE05127M
Mouse soluble receptor actiVator of nuclear factor-kB ligand sRANKL ELISA Kit from CUSABIO Detection Range 12 5 pg/ml-800 pg/ml Sensitivity 3 12 pg/ml Sample serum plasma tissue homogenates Method Sandwich