Learn More
American Research Products Inc Mouse soluble Triggering Recep
Catalog No. 5000328315
Encompass
Mouse soluble Triggering Receptor EXpresses on Myeloid Cells-1 sTREM-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28315 Supplier American Research Products Inc Supplier No. CSBE13848M
Mouse soluble Triggering Receptor EXpresses on Myeloid Cells-1 sTREM-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich