Learn More
Biomatik Corporation Mouse Solute Carrier Family 30, Member 5 (SLC30A5) ELISA Kit
Supplier:  Biomatik Corporation EKU07405
Encompass_Preferred
Mouse Solute Carrier Family 30, Member 5 (SLC30A5) ELISA Kit for detecting SLC30A5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.108ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R4H9. Gene ID: 69048. Target Synonyms: ZTL1; ZnT-5; ZNT5; ZNTL1; Zinc Transporter 5; ZnT-like transporter 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8753