Learn More
Invitrogen™ Mouse Somatostatin Competitive ELISA Kit
Description
The Mouse Somatostatin (SST) ELISA quantitates SST in serum, plasma and other biological fluids. Principle of the method The Competitive mouse ELISA research use only kit is designed to quantitatively measure Somatostatin (SST) in mouse. A Somatostatin standard is provided to generate a standard curve for the assay and all samples are read off a user generated standard curve. Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to Somatostatin is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of Somatostatin in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Specifications
Specifications
| Accession Number | P60041 |
| Assay Range | 7.81-500 pg/mL |
| Assay Sensitivity | 4.69 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Cell Lysates, Plasma, Serum, Tissue Homogenates |
| Target Species | Mouse |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.