Learn More
American Research Products Inc Mouse Sonic hedgehog protein
Catalog No. 5000327169
Encompass
Mouse Sonic hedgehog protein (SHH) ELISA kit from CUSABIO Detection Range 28 pg/mL-1800 pg/mL Sensitivity 7 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27169 Supplier American Research Products Inc Supplier No. CSBEL021266MO
Mouse Sonic hedgehog protein (SHH) ELISA kit from CUSABIO Detection Range 28 pg/mL-1800 pg/mL Sensitivity 7 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich