Learn More
Abcam Mouse SOST ELISA Kit, 1 x 96 Tests
Catalog No. 502660611
Encompass_Preferred
Mouse SOST ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse SOST in Mouse in Cell culture media, EDTA Plasma, Citrate plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-0611 Supplier Abcam Supplier No. AB3091141X96TESTS
Mouse SOST ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse SOST in Mouse in Cell culture media, EDTA Plasma, Citrate plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement