Learn More
Sino Biological Recombinant Mouse SOX2 Protein (His Tag)
Supplier:  Sino Biological 57011M07E100
Encompass_Preferred
A DNA sequence encoding the Mouse SOX2 (P48432) (Met1-Met319) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-2958