Learn More
American Research Products Inc Mouse SPARC like protein 1 SP
Catalog No. 5000327599
Encompass
Mouse SPARC-like protein 1 (SPARCL1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27599 Supplier American Research Products Inc Supplier No. CSBEL022487MO
Mouse SPARC-like protein 1 (SPARCL1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich