Learn More
Sino Biological Anti-SPARC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50494RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Osteonectin / SPARC (rM Osteonectin / SPARC; Catalog#50494-M08H; NP_033268.1; Met 1-Ile 302). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2105