Learn More
Abcam Mouse specific HRP/DAB Detection IHC Kit, 15ML
Catalog No. NC0792039
Encompass_Preferred
Mouse specific HRP/DAB IHC Detection Kit ab64259 is a complete kit to detect a primary antibody raised in mouse, using HRP-labeled-streptavidin and a biotinylated anti-mouse secondary antibody.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0792039 Supplier Abcam Supplier No. AB6425915ML
Mouse specific HRP/DAB IHC Detection Kit ab64259 is a complete kit to detect a primary antibody raised in mouse, using HRP-labeled-streptavidin and a biotinylated anti-mouse secondary antibody.
The product is subject to the following: Abcam Restricted Use Statement