Learn More
Biomatik Corporation Mouse Spermidine/Spermine N1-Acetyltransferase 1 (SAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU07425
Encompass_Preferred
Mouse Spermidine/Spermine N1-Acetyltransferase 1 (SAT1) ELISA Kit for detecting SAT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48026. Gene ID: 20229. Target Synonyms: DC21; KFSD; SAT; SSAT; SSAT-1; Diamine Acetyltransferase 1; Polyamine N-acetyltransferase 1; Putrescine acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5687