Learn More
ACROBiosystems Mouse Spike RBD 100ug
Supplier:  ACROBiosystems SPDS298100UG
Encompass_Preferred
Its been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits S1 and S2. S1 mainly contains a receptor binding domain (RBD) which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses as well as protective immunity.
Catalog No. 50-253-4145