Learn More
ACROBiosystems Anti-SARS-CoV-2 Spike RBD Antibody, Mouse IgG1 (AM334) (Beta & Gamma Specific) (MALS verified)
Catalog No. 502718889
Encompass_Preferred
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
Catalog No. 50-271-8889 Supplier ACROBiosystems Supplier No. SPDM3341MG
Add to Cart
Add to Cart
Request a Quote
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.