Learn More
World Precision Instrument Mouse Spinal Adaptor
SDP
Catalog No. 502539136
Encompass
The design was inspired by the need for a practical, versatile, and economical instrument for stabilizing the spinal column. The Spinal Adaptors are accessories to be mounted on the 505211, 505351 or other brands of stereotaxic instruments, converting them for precise spinal surgery applications.
Catalog No. 50-253-9136 Supplier World Precision Instrument Supplier No. 505289
The design was inspired by the need for a practical, versatile, and economical instrument for stabilizing the spinal column. The Spinal Adaptors are accessories to be mounted on the 505211, 505351 or other brands of stereotaxic instruments, converting them for precise spinal surgery applications.