Learn More
Sino Biological Mouse SPINT2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50042RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SPINT2 / Placental bikunin (rM SPINT2 / Placental bikunin; Catalog#50042-M08H; NP_001076017.1; Met1-Lys140). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3938