Learn More
iQ Biosciences C57BL/6 Mouse Splenocytes, 25M cells/vial
Catalog No. NC1671489
Encompass_Preferred
High-quality, high-viability (typically > 70%) splenocytes from the C57BL/6 mouse, the most commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1671489 Supplier iQ Biosciences Supplier No. IQBMSP202
High-quality, high-viability (typically > 70%) splenocytes from the C57BL/6 mouse, the most commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.