Learn More
American Research Products Inc Mouse Stem cell factor mast ce
Catalog No. 5000326544
Encompass
Mouse Stem cell factor/mast cell growth factor SCF/MGF ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26544 Supplier American Research Products Inc Supplier No. CSBE04719M
Mouse Stem cell factor/mast cell growth factor SCF/MGF ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich