Learn More
Bioworld MOUSE STEM CELL FACTOR (REC
Supplier:  Bioworld 220605811
Encompass_Preferred
MOUSE STEM CELL FACTOR (RECOMBINANT): Recombinant Murine Stem Cell Factor (SCF) Source: E.Coli Description: Recombinant murine SCF produced in E.Coli is a single, non-glycosylated polypeptide chain containing 164 amino acids and havi...
Catalog No. 50-253-681