Learn More
Biomatik Corporation Mouse Stem Cell Factor Receptor (SCFR) ELISA Kit
Supplier:  Biomatik Corporation EKU07468
Encompass_Preferred
Mouse Stem Cell Factor Receptor (SCFR) ELISA Kit for detecting SCFR in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05532. Gene ID: 16590. Target Synonyms: CD117; KIT; C-Kit; PBT; p145 c-Kit; C-Kit Receptor; Tyrosine-Protein Kinase; V-Kit Hardy-Zuckerman 4 Feline Sarcoma Viral Oncogene Homolog; Piebald Trait. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6300