Learn More
Abcam Mouse sTNF RII ELISA Kit (TNFRSF1B), 1 x 96 Tests
Catalog No. 502657655
Encompass_Preferred
Mouse sTNF RII ELISA Kit (TNFRSF1B) is a single-wash 90-min Simplestep used to quantify Mouse sTNF RII (TNFRSF1B) with a sensitivity of 1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7655 Supplier Abcam Supplier No. AB2024121X96TESTS
Mouse sTNF RII ELISA Kit (TNFRSF1B) is a single-wash 90-min Simplestep used to quantify Mouse sTNF RII (TNFRSF1B) with a sensitivity of 1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement