Learn More
ATCC Mouse STR Profiling Service
Catalog No. 502382224
Encompass
FTA Sample Collection Kit for Mouse Cell Authentication Service utilizes short tandem repeat (STR) profiling. Trained experts amplify 18 STR loci plus 2 specific loci for detection of human or African green monkey species within the submitted sample. A comprehensive analysis report interprets both karyotypically normal and abnormal cell lines, includes an electropherogram supporting the allele calls at each locus, known reference profiling against the ATCC Mouse STR database and a comprehensive interpretation of results. Sample kit mailed upon purchase. Download the instructions for sample preparation and submission. This service is performed according to Almeida JL, et al. Interlaboratory study to validate a STR profiling method for intraspecies identification of mouse cell lines. PLoS ONE 14(6): e0218412, 2019. PubMed: 31220119.
Catalog No. 50-238-2224 Supplier ATCC Supplier No. 137XV
FTA Sample Collection Kit for Mouse Cell Authentication Service utilizes short tandem repeat (STR) profiling. Trained experts amplify 18 STR loci plus 2 specific loci for detection of human or African green monkey species within the submitted sample. A comprehensive analysis report interprets both karyotypically normal and abnormal cell lines, includes an electropherogram supporting the allele calls at each locus, known reference profiling against the ATCC Mouse STR database and a comprehensive interpretation of results. Sample kit mailed upon purchase. Download the instructions for sample preparation and submission. This service is performed according to Almeida JL, et al. Interlaboratory study to validate a STR profiling method for intraspecies identification of mouse cell lines. PLoS ONE 14(6): e0218412, 2019. PubMed: 31220119.