Learn More
American Research Products Inc Mouse Stromal cell deriVed fac
Catalog No. 5000329069
Encompass
Mouse Stromal cell deriVed factor 1b SDF-1b ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29069 Supplier American Research Products Inc Supplier No. CSBE04723M
Mouse Stromal cell deriVed factor 1b SDF-1b ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich