Learn More
American Research Products Inc Mouse stromal cell deriVed fac
Catalog No. 5000327881
Encompass
Mouse stromal cell deriVed factor 1a (SDF-1a/SDF1A) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27881 Supplier American Research Products Inc Supplier No. CSBEQ027494MO
Mouse stromal cell deriVed factor 1a (SDF-1a/SDF1A) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich