Learn More
Biomatik Corporation Mouse Stromal Cell Derived Factor 1 (SDF1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11252
Encompass_Preferred
Mouse Stromal Cell Derived Factor 1 (SDF1) Mini sample ELISA Kit for detecting SDF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 25.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40224. Gene ID: 20315. Target Synonyms: CXCL12; PBSF; P-BSF; SCYB12; SDF1-A; SDF1-B; SDF1A; SDF1B; TLSF-A; TLSF-B; TPAR1; Pre-B Cell Growth-Stimulating Factor; Chemokine C-X-C-Motif Ligand 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8184