Learn More
Sino Biological Anti-Syntaxin 6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106430T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the mouse Syntaxin 6/STX6, and purified by antigen affinity chromatography.
Catalog No. 50-241-0721