Learn More
Biomatik Corporation Mouse Succinate Dehydrogenase Complex Subunit C (SDHC) ELISA Kit
Supplier:  Biomatik Corporation EKU07489
Encompass_Preferred
Mouse Succinate Dehydrogenase Complex Subunit C (SDHC) ELISA Kit for detecting SDHC in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.071ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CZB0. Gene ID: 66052. Target Synonyms: PGL3; CYB560, SDH3; CYBL; QPs1; Integral membrane protein CII-3; Succinate-ubiquinone oxidoreductase cytochrome B large subunit; Succinate dehydrogenase cytochrome b560. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8748