Learn More
Biomatik Corporation Mouse Succinate Dehydrogenase Complex Subunit B (SDHB) ELISA Kit
Supplier:  Biomatik Corporation EKU07488
Encompass_Preferred
Mouse Succinate Dehydrogenase Complex Subunit B (SDHB) ELISA Kit for detecting SDHB in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CQA3. Gene ID: 67680. Target Synonyms: SDH1; SDH; Ip; Iron Sulfur; Iron-sulfur subunit of complex II; Succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5859