Learn More
Biomatik Corporation Mouse Sulfotransferase 1A1 (SULT1A1) ELISA Kit
Supplier:  Biomatik Corporation EKU06577
Encompass_Preferred
Mouse Sulfotransferase 1A1 (SULT1A1) ELISA Kit for detecting SULT1A1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52840. Target Synonyms: PST; HAST1/HAST2; ST1A3; STP1; TSPST1; Phenol Sulfotransferase; Aryl sulfotransferase 1; Phenol sulfotransferase 1; Thermostable phenol sulfotransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6638