Learn More
American Research Products Inc Mouse SuperoXide dismutase Mn
Catalog No. 5000328182
Encompass
Mouse SuperoXide dismutase [Mn] mitochondrial (SOD2) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28182 Supplier American Research Products Inc Supplier No. CSBEL022398MO
Mouse SuperoXide dismutase [Mn] mitochondrial (SOD2) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich