Learn More
American Research Products Inc Mouse suppressors of cytokine
Catalog No. 5000329349
Encompass
Mouse suppressors of cytokine signaling 1 (SOCS-1) ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29349 Supplier American Research Products Inc Supplier No. CSBE17346M
Mouse suppressors of cytokine signaling 1 (SOCS-1) ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich